Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Fmoc-L-glutamic acid 1-benzyl ester, 95%, Thermo Scientific Chemicals

Catalog Number AAH6655103
Click to view available options
Quantity:
1 g
5 g
122350-52-1
C27H25NO6
459.498
MFCD00080272
FMWLYDDRYGOYMY-DEOSSOPVSA-N
fmoc-glu-obzl, s-4-9h-fluoren-9-yl methoxy carbonyl amino-5-benzyloxy-5-oxopentanoic acid, 4s-5-benzyloxy-4-9h-fluoren-9-ylmethoxy carbonyl amino-5-oxopentanoic acid, fmoc-l-glutamic acid alpha-benzyl ester, nalpha-fmoc-l-glutamic acid alpha-benzyl ester
10049854
(4S)-4-(9H-fluoren-9-ylmethoxycarbonylamino)-5-oxo-5-phenylmethoxypentanoic acid
C1=CC=C(C=C1)COC(=O)C(CCC(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
122350-52-1
C27H25NO6
459.498
MFCD00080272
FMWLYDDRYGOYMY-DEOSSOPVSA-N
fmoc-glu-obzl, s-4-9h-fluoren-9-yl methoxy carbonyl amino-5-benzyloxy-5-oxopentanoic acid, 4s-5-benzyloxy-4-9h-fluoren-9-ylmethoxy carbonyl amino-5-oxopentanoic acid, fmoc-l-glutamic acid alpha-benzyl ester, nalpha-fmoc-l-glutamic acid alpha-benzyl ester
10049854
(4S)-4-(9H-fluoren-9-ylmethoxycarbonylamino)-5-oxo-5-phenylmethoxypentanoic acid
C1=CC=C(C=C1)COC(=O)C(CCC(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Formula Weight 459.5
Percent Purity 95%
Quantity 1 g
Physical Form Powder
Chemical Name or Material N-Fmoc-L-glutamic acid 1-benzyl ester
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit