Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Fmoc-L-glutamic acid 5-cyclohexyl ester, 98%

Click to view available options
Quantity:
5g
25g
150047-85-1
C26H29NO6
451.519
MFCD00270538
ODIPEUXZAYGDMU-QHCPKHFHSA-N
fmoc-glu ochx-oh,fmoc-glu ochex-oh,s-2-9h-fluoren-9-yl methoxy carbonyl amino-5-cyclohexyloxy-5-oxopentanoic acid,ambotzfaa1719,n-alpha-9-fluorenylmethyloxycarbonyl-l-glutamic-acid-gamma-cyclohexyl ester,2s-5-cyclohexyloxy-2-9h-fluoren-9-yl methoxy carbonyl amino-5-oxopentanoic acid non-preferred name,2s-5-cyclohexyloxy-2-9h-fluoren-9-ylmethoxy carbonyl amino-5-oxopentanoic acid
51340555
(2S)-5-cyclohexyloxy-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-oxopentanoic acid
C1CCC(CC1)OC(=O)CCC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
150047-85-1
C26H29NO6
451.519
MFCD00270538
ODIPEUXZAYGDMU-QHCPKHFHSA-N
fmoc-glu ochx-oh,fmoc-glu ochex-oh,s-2-9h-fluoren-9-yl methoxy carbonyl amino-5-cyclohexyloxy-5-oxopentanoic acid,ambotzfaa1719,n-alpha-9-fluorenylmethyloxycarbonyl-l-glutamic-acid-gamma-cyclohexyl ester,2s-5-cyclohexyloxy-2-9h-fluoren-9-yl methoxy carbonyl amino-5-oxopentanoic acid non-preferred name,2s-5-cyclohexyloxy-2-9h-fluoren-9-ylmethoxy carbonyl amino-5-oxopentanoic acid
51340555
(2S)-5-cyclohexyloxy-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-oxopentanoic acid
C1CCC(CC1)OC(=O)CCC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
Formula Weight 451.52
Physical Form Solid
Percent Purity 98%
Quantity 25g
Chemical Name or Material N-Fmoc-L-glutamic acid 5-cyclohexyl ester

storageNote1 : -30° to -10°C