Alfa Aesar™ N-Fmoc-L-homophenylalanine, 95%

Manufacturer: Alfa Aesar™ H5197303

Quantity 1g

 View more versions of this product

Catalog No. AAH5197303

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 132684-59-4
    Molecular Formula C25H23NO4
    Formula Weight 401.46
    MDL Number MFCD00077046
    Beilstein 4847669
    Synonym (S)-2-(Fmoc-amino)-4-phenylbutyric acid; Fmoc-Homophe-OH
    Chemical Name or Material N-Fmoc-L-homophenylalanine

    Specifications

    Formula Weight 401.46
    Molecular Formula C25H23NO4
    MDL Number MFCD00077046
    CAS 132684-59-4
    Beilstein 4847669
    Quantity 1g
    Synonym (S)-2-(Fmoc-amino)-4-phenylbutyric acid; Fmoc-Homophe-OH
    Chemical Name or Material N-Fmoc-L-homophenylalanine
    Assay 95%

    Description

    Applications
    It is used as an active pharmaceutical intermediate.

    Solubility
    Slightly soluble in water.

    Notes
    Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit