Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Fmoc-L-norleucine, 98%

Click to view available options
Quantity:
1g
5g
25g
77284-32-3
C21H23NO4
353.418
MFCD00037537
VCFCFPNRQDANPN-IBGZPJMESA-N
fmoc-nle-oh,fmoc-l-norleucine,n-fmoc-l-norleucine,s-2-fmoc-amino caproic acid,s-2-9h-fluoren-9-yl methoxy carbonyl amino hexanoic acid,s-2-9h-fluoren-9-ylmethoxycarbonylamino-hexanoic acid,2s-2-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid,fmoc-norleucine,fmocnleoh,fmoc-nle
7009636
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)hexanoic acid
CCCCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
77284-32-3
C21H23NO4
353.418
MFCD00037537
VCFCFPNRQDANPN-IBGZPJMESA-N
fmoc-nle-oh,fmoc-l-norleucine,n-fmoc-l-norleucine,s-2-fmoc-amino caproic acid,s-2-9h-fluoren-9-yl methoxy carbonyl amino hexanoic acid,s-2-9h-fluoren-9-ylmethoxycarbonylamino-hexanoic acid,2s-2-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid,fmoc-norleucine,fmocnleoh,fmoc-nle
7009636
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)hexanoic acid
CCCCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
Melting Point 140°C to 142°C
Quantity 5g
Formula Weight 353.42
Percent Purity 98%
Chemical Name or Material N-Fmoc-L-norleucine

Recommended Storage : 2° to 8°C