Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Fmoc-L-phenylalanine pentafluorophenyl ester, 96%

Click to view available options
Quantity:
250mg
1g
5g
86060-92-6
C30H20F5NO4
553.485
MFCD00065669
WKHPSOMXNCTXPK-QFIPXVFZSA-N
fmoc-phe-opfp,n-alpha-9-fluorenylmethyloxycarbonyl-l-phenylalanine pentafluorphenyl ester,2,3,4,5,6-pentafluorophenyl 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-phenylpropanoate,fmoc-phe-opfp,l-phenylalanine,n-9h-fluoren-9-ylmethoxy carbonyl-, 2,3,4,5,6-pentafluorophenyl ester,fmoc-l-phenylalanine pentafluorophenyl ester,n-fmoc-l-phenylalanine pentafluorophenyl ester,n-9-fluorenylmethoxycarbonyl-l-phenylalanine pentafluorophenyl ester,n-9h-fluorene-9-ylmethoxycarbonyl-l-phenylalanine pentafluorophenyl ester,pentafluorophenyl n-9h-fluoren-9-ylmethoxy carbonyl-l-phenylalaninate
2733280
(2,3,4,5,6-pentafluorophenyl) (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-phenylpropanoate
C1=CC=C(C=C1)CC(C(=O)OC2=C(C(=C(C(=C2F)F)F)F)F)NC(=O)OCC3C4=CC=CC=C4C5=CC=CC=C35
86060-92-6
C30H20F5NO4
553.485
MFCD00065669
WKHPSOMXNCTXPK-QFIPXVFZSA-N
fmoc-phe-opfp,n-alpha-9-fluorenylmethyloxycarbonyl-l-phenylalanine pentafluorphenyl ester,2,3,4,5,6-pentafluorophenyl 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-phenylpropanoate,fmoc-phe-opfp,l-phenylalanine,n-9h-fluoren-9-ylmethoxy carbonyl-, 2,3,4,5,6-pentafluorophenyl ester,fmoc-l-phenylalanine pentafluorophenyl ester,n-fmoc-l-phenylalanine pentafluorophenyl ester,n-9-fluorenylmethoxycarbonyl-l-phenylalanine pentafluorophenyl ester,n-9h-fluorene-9-ylmethoxycarbonyl-l-phenylalanine pentafluorophenyl ester,pentafluorophenyl n-9h-fluoren-9-ylmethoxy carbonyl-l-phenylalaninate
2733280
(2,3,4,5,6-pentafluorophenyl) (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-phenylpropanoate
C1=CC=C(C=C1)CC(C(=O)OC2=C(C(=C(C(=C2F)F)F)F)F)NC(=O)OCC3C4=CC=CC=C4C5=CC=CC=C35
Beilstein 4241247
Formula Weight 553.49
Physical Form Powder
Percent Purity 96%
Color White
Melting Point 149°C to 153°C
Quantity 5g
Chemical Name or Material N-Fmoc-L-phenylalanine pentafluorophenyl ester

storageNote1 : 2° to 8°C