Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Fmoc-L-phenylalaninol, 95%

Click to view available options
Quantity:
5g
25g
129397-83-7
C24H23NO3
373.452
MFCD00235941
SJGBJASOHDROCR-SFHVURJKSA-N
fmoc-phenylalaninol,fmoc-l-phenylalaninol,s-9h-fluoren-9-yl methyl 1-hydroxy-3-phenylpropan-2-yl carbamate,fmoc-phe-ol,s-9h-fluoren-9-yl methyl 1-hydroxy-3-phenylpropan-2-ylcarbamate,n-fmoc-l-phenylalaninol,s-2-9-fluorenylmethyloxycarbonyl amino-3-phenyl-1-propanol,9h-fluoren-9-ylmethyl n-2s-1-hydroxy-3-phenylpropan-2-yl carbamate,pubchem12466,carbamic acid,n-1s-1-hydroxymethyl-2-phenylethyl-, 9h-fluoren-9-ylmethyl ester
7019531
9H-fluoren-9-ylmethyl N-[(2S)-1-hydroxy-3-phenylpropan-2-yl]carbamate
C1=CC=C(C=C1)CC(CO)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
129397-83-7
C24H23NO3
373.452
MFCD00235941
SJGBJASOHDROCR-SFHVURJKSA-N
fmoc-phenylalaninol,fmoc-l-phenylalaninol,s-9h-fluoren-9-yl methyl 1-hydroxy-3-phenylpropan-2-yl carbamate,fmoc-phe-ol,s-9h-fluoren-9-yl methyl 1-hydroxy-3-phenylpropan-2-ylcarbamate,n-fmoc-l-phenylalaninol,s-2-9-fluorenylmethyloxycarbonyl amino-3-phenyl-1-propanol,9h-fluoren-9-ylmethyl n-2s-1-hydroxy-3-phenylpropan-2-yl carbamate,pubchem12466,carbamic acid,n-1s-1-hydroxymethyl-2-phenylethyl-, 9h-fluoren-9-ylmethyl ester
7019531
9H-fluoren-9-ylmethyl N-[(2S)-1-hydroxy-3-phenylpropan-2-yl]carbamate
C1=CC=C(C=C1)CC(CO)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
Quantity 25g
Formula Weight 373.45
Physical Form Solid
Percent Purity 95%
Chemical Name or Material N-Fmoc-L-phenylalaninol

storageNote1 : -30° to -10°C