Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Fmoc-L-proline N-succinimidyl ester, 95%

Click to view available options
Quantity:
5g
25g
109074-94-4
C24H22N2O6
434.448
MFCD00065673
JZBHVBQPWXZALQ-FQEVSTJZSA-N
fmoc-pro-osu,2-2,5-dioxopyrrolidin-1-yl 1-9h-fluoren-9-ylmethyl 2s-pyrrolidine-1,2-dicarboxylate,ambotzfaa6500,n-alpha-9-fluorenylmethyloxycarbonyl-l-proline succinimidyl ester,2,5-dioxo-1-pyrrolidinyl 1-9h-fluoren-9-ylmethoxy carbonyl-l-prolinate,2,5-dioxopyrrolidin-1-yl 1-9h-fluoren-9-yl methoxy carbonyl-l-prolinate
51340599
2-O-(2,5-dioxopyrrolidin-1-yl) 1-O-(9H-fluoren-9-ylmethyl) (2S)-pyrrolidine-1,2-dicarboxylate
C1CC(N(C1)C(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24)C(=O)ON5C(=O)CCC5=O
109074-94-4
C24H22N2O6
434.448
MFCD00065673
JZBHVBQPWXZALQ-FQEVSTJZSA-N
fmoc-pro-osu,2-2,5-dioxopyrrolidin-1-yl 1-9h-fluoren-9-ylmethyl 2s-pyrrolidine-1,2-dicarboxylate,ambotzfaa6500,n-alpha-9-fluorenylmethyloxycarbonyl-l-proline succinimidyl ester,2,5-dioxo-1-pyrrolidinyl 1-9h-fluoren-9-ylmethoxy carbonyl-l-prolinate,2,5-dioxopyrrolidin-1-yl 1-9h-fluoren-9-yl methoxy carbonyl-l-prolinate
51340599
2-O-(2,5-dioxopyrrolidin-1-yl) 1-O-(9H-fluoren-9-ylmethyl) (2S)-pyrrolidine-1,2-dicarboxylate
C1CC(N(C1)C(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24)C(=O)ON5C(=O)CCC5=O
Formula Weight 434.45
Physical Form Solid
Percent Purity 95%
Quantity 25g
Chemical Name or Material N-Fmoc-L-proline N-succinimidyl ester

storageNote1 : -30° to -10°C