Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Fmoc-L-proline N-succinimidyl ester, 95%, Thermo Scientific™

Catalog Number AAH6650706
Click to view available options
Quantity:
5 g
25 g
109074-94-4
C24H22N2O6
434.448
MFCD00065673
JZBHVBQPWXZALQ-FQEVSTJZSA-N
fmoc-pro-osu, 2-2,5-dioxopyrrolidin-1-yl 1-9h-fluoren-9-ylmethyl 2s-pyrrolidine-1,2-dicarboxylate, ambotzfaa6500, n-alpha-9-fluorenylmethyloxycarbonyl-l-proline succinimidyl ester, 2,5-dioxo-1-pyrrolidinyl 1-9h-fluoren-9-ylmethoxy carbonyl-l-prolinate, 2,5-dioxopyrrolidin-1-yl 1-9h-fluoren-9-yl methoxy carbonyl-l-prolinate
51340599
2-O-(2,5-dioxopyrrolidin-1-yl) 1-O-(9H-fluoren-9-ylmethyl) (2S)-pyrrolidine-1,2-dicarboxylate
C1CC(N(C1)C(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24)C(=O)ON5C(=O)CCC5=O
109074-94-4
C24H22N2O6
434.448
MFCD00065673
JZBHVBQPWXZALQ-FQEVSTJZSA-N
fmoc-pro-osu, 2-2,5-dioxopyrrolidin-1-yl 1-9h-fluoren-9-ylmethyl 2s-pyrrolidine-1,2-dicarboxylate, ambotzfaa6500, n-alpha-9-fluorenylmethyloxycarbonyl-l-proline succinimidyl ester, 2,5-dioxo-1-pyrrolidinyl 1-9h-fluoren-9-ylmethoxy carbonyl-l-prolinate, 2,5-dioxopyrrolidin-1-yl 1-9h-fluoren-9-yl methoxy carbonyl-l-prolinate
51340599
2-O-(2,5-dioxopyrrolidin-1-yl) 1-O-(9H-fluoren-9-ylmethyl) (2S)-pyrrolidine-1,2-dicarboxylate
C1CC(N(C1)C(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24)C(=O)ON5C(=O)CCC5=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Formula Weight 434.45
Percent Purity 95%
Quantity 5 g
Physical Form Solid
Chemical Name or Material N-Fmoc-L-proline N-succinimidyl ester
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit