Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Fmoc-L-threonine, 98%

Click to view available options
Quantity:
1g
5g
118609-38-4
C19H21NO6
359.378
MFCD00153365
CGNQPFAECJFQNV-VFZPIINCSA-N
2r,3s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-hydroxybutanoic acid hydrate,2r,3s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-hydroxybutanoic acid hydrate,fmoc-d-thr-oh.h2o,n-alpha-9-fluorenylmethyloxycarbonyl-d-threonine monohydrate,n-9h-fluoren-9-yl methoxy carbonyl-d-threonine-water 1/1
71607443
(2R,3S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-hydroxybutanoic acid;hydrate
CC(C(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13)O.O
118609-38-4
C19H21NO6
359.378
MFCD00153365
CGNQPFAECJFQNV-VFZPIINCSA-N
2r,3s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-hydroxybutanoic acid hydrate,2r,3s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-hydroxybutanoic acid hydrate,fmoc-d-thr-oh.h2o,n-alpha-9-fluorenylmethyloxycarbonyl-d-threonine monohydrate,n-9h-fluoren-9-yl methoxy carbonyl-d-threonine-water 1/1
71607443
(2R,3S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-hydroxybutanoic acid;hydrate
CC(C(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13)O.O
Applications
It is employed as a intermediate for pharmaceutical.

Solubility
Sparingly soluble in water (0.053 g/L) (25°C).

Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Formula Weight 341.36
Physical Form Solid
Percent Purity 98%
Quantity 1g
Chemical Name or Material N-Fmoc-L-threonine

storageNote1 : 2° to 8°C