Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Fmoc-L-threoninol, 98%

Click to view available options
Quantity:
250mg
1g
5g
176380-53-3
C19H21NO4
327.38
MFCD00270218
YOKDHMTZJSRRIQ-KZULUSFZSA-N
fmoc-threoninol,n-fmoc-l-threonol,fmoc-thr-ol,fmoc-l-threoninol,n-fmoc-l-threoninol,9h-fluoren-9-yl methyl 2r,3r-1,3-dihydroxybutan-2-yl carbamate,1r,2r-2-hydroxy-1-hydroxymethylpropyl carbamic acid 9h-fluoren-9-ylmethyl ester,9-fluorenylmethyloxycarbonyl-l-threoninol,9h-fluoren-9-ylmethyl n-2r,3r-1,3-dihydroxybutan-2-yl carbamate,fmoc-l-threonol
7020321
9H-fluoren-9-ylmethyl N-[(2R,3R)-1,3-dihydroxybutan-2-yl]carbamate
CC(C(CO)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13)O
176380-53-3
C19H21NO4
327.38
MFCD00270218
YOKDHMTZJSRRIQ-KZULUSFZSA-N
fmoc-threoninol,n-fmoc-l-threonol,fmoc-thr-ol,fmoc-l-threoninol,n-fmoc-l-threoninol,9h-fluoren-9-yl methyl 2r,3r-1,3-dihydroxybutan-2-yl carbamate,1r,2r-2-hydroxy-1-hydroxymethylpropyl carbamic acid 9h-fluoren-9-ylmethyl ester,9-fluorenylmethyloxycarbonyl-l-threoninol,9h-fluoren-9-ylmethyl n-2r,3r-1,3-dihydroxybutan-2-yl carbamate,fmoc-l-threonol
7020321
9H-fluoren-9-ylmethyl N-[(2R,3R)-1,3-dihydroxybutan-2-yl]carbamate
CC(C(CO)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13)O
Quantity 250mg
Formula Weight 327.38
Physical Form Solid
Percent Purity 98%
Chemical Name or Material N-Fmoc-L-threoninol

storageNote1 : -30° to -10°C