Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Fmoc-L-valine N-succinimidyl ester, 95%

Click to view available options
Quantity:
5g
25g
130878-68-1
C24H24N2O6
436.464
MFCD00153370
JPJMNCROLRPFHI-QFIPXVFZSA-N
fmoc-val-osu,s-2,5-dioxopyrrolidin-1-yl 2-9h-fluoren-9-yl methoxy carbonyl amino-3-methylbutanoate,n-fmoc-l-valine n-succinimidyl ester,fmoc-l-valine-osu,2,5-dioxopyrrolidin-1-yl 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-methylbutanoate,fmoc-l-val-osu,ambotzfaa6560,fmoc-l-valine n-hydroxysuccinimide ester,n-9h-fluorene-9-ylmethoxycarbonyl-l-valine succinimidyl ester,n-alpha-9-fluorenylmethyloxycarbonyl-l-valine succinimidyl ester
11339559
(2,5-dioxopyrrolidin-1-yl) (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-methylbutanoate
CC(C)C(C(=O)ON1C(=O)CCC1=O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
130878-68-1
C24H24N2O6
436.464
MFCD00153370
JPJMNCROLRPFHI-QFIPXVFZSA-N
fmoc-val-osu,s-2,5-dioxopyrrolidin-1-yl 2-9h-fluoren-9-yl methoxy carbonyl amino-3-methylbutanoate,n-fmoc-l-valine n-succinimidyl ester,fmoc-l-valine-osu,2,5-dioxopyrrolidin-1-yl 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-methylbutanoate,fmoc-l-val-osu,ambotzfaa6560,fmoc-l-valine n-hydroxysuccinimide ester,n-9h-fluorene-9-ylmethoxycarbonyl-l-valine succinimidyl ester,n-alpha-9-fluorenylmethyloxycarbonyl-l-valine succinimidyl ester
11339559
(2,5-dioxopyrrolidin-1-yl) (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-methylbutanoate
CC(C)C(C(=O)ON1C(=O)CCC1=O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
Applications
N-Fmoc-L-valine N-succinimidyl ester is useful for solid phase peptide synthesis techniques.

Solubility
Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Formula Weight 436.46
Physical Form Powder
Percent Purity 95%
Color White
Quantity 25g
Chemical Name or Material N-Fmoc-L-valine N-succinimidyl ester

storageNote1 : 2° to 8°C