Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Fmoc-L-valine pentafluorophenyl ester, 98%

Click to view available options
Quantity:
1g
5g
86060-87-9
C26H20F5NO4
505.441
MFCD00037650
TZEGAVSWQUEHAQ-QHCPKHFHSA-N
fmoc-val-opfp,n-alpha-9-fluorenylmethyloxycarbonyl-l-valine pentafluorphenyl ester,l-valine, n-9h-fluoren-9-ylmethoxy carbonyl-, pentafluorophenyl ester,l-valine,n-9h-fluoren-9-ylmethoxy carbonyl-, 2,3,4,5,6-pentafluorophenyl ester,fmoc-l-valine pentafluorophenyl ester,fmoc-val-opfp hplc,n-9h-fluorene-9-ylmethoxycarbonyl-l-valine pentafluorophenyl ester,n-9-fluorenylmethoxycarbonyl-l-valine pentafluorophenyl ester,2,3,4,5,6-pentafluorophenyl 2s-2-9h-fluoren-9-ylmethoxycarbonylamino-3-methylbutanoate,2,3,4,5,6-pentafluorophenyl 2s-2-fluoren-9-ylmethoxy carbonylamino-3-meth ylbutanoate
2733281
(2,3,4,5,6-pentafluorophenyl) (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-methylbutanoate
CC(C)C(C(=O)OC1=C(C(=C(C(=C1F)F)F)F)F)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
86060-87-9
C26H20F5NO4
505.441
MFCD00037650
TZEGAVSWQUEHAQ-QHCPKHFHSA-N
fmoc-val-opfp,n-alpha-9-fluorenylmethyloxycarbonyl-l-valine pentafluorphenyl ester,l-valine, n-9h-fluoren-9-ylmethoxy carbonyl-, pentafluorophenyl ester,l-valine,n-9h-fluoren-9-ylmethoxy carbonyl-, 2,3,4,5,6-pentafluorophenyl ester,fmoc-l-valine pentafluorophenyl ester,fmoc-val-opfp hplc,n-9h-fluorene-9-ylmethoxycarbonyl-l-valine pentafluorophenyl ester,n-9-fluorenylmethoxycarbonyl-l-valine pentafluorophenyl ester,2,3,4,5,6-pentafluorophenyl 2s-2-9h-fluoren-9-ylmethoxycarbonylamino-3-methylbutanoate,2,3,4,5,6-pentafluorophenyl 2s-2-fluoren-9-ylmethoxy carbonylamino-3-meth ylbutanoate
2733281
(2,3,4,5,6-pentafluorophenyl) (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-methylbutanoate
CC(C)C(C(=O)OC1=C(C(=C(C(=C1F)F)F)F)F)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
Beilstein 3640481
Formula Weight 505.44
Physical Form Solid
Percent Purity 98%
Quantity 5g
Chemical Name or Material N-Fmoc-L-valine pentafluorophenyl ester