Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Fmoc-N-methyl-D-valine, 95%

Click to view available options
Quantity:
250mg
1g
5g
103478-58-6
C21H23NO4
353.418
MFCD00153396
YCXXXPZNQXXRIG-LJQANCHMSA-N
fmoc-n-methyl-d-valine,fmoc-d-n-me-val-oh,fmoc-n-me-d-val-oh,fmoc-nalpha-methyl-d-valine,r-2-9h-fluoren-9-yl methoxy carbonyl methyl amino-3-methylbutanoic acid,2r-2-9h-fluoren-9-ylmethoxy carbonyl methyl amino-3-methylbutanoic acid,ambotzfaa1533,fmoc-d-n-me-ala-oh,fmoc-d-n-me-val-oh,n-fmoc-n-methyl-d-valine
16213159
(2R)-2-[9H-fluoren-9-ylmethoxycarbonyl(methyl)amino]-3-methylbutanoic acid
CC(C)C(C(=O)O)N(C)C(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
103478-58-6
C21H23NO4
353.418
MFCD00153396
YCXXXPZNQXXRIG-LJQANCHMSA-N
fmoc-n-methyl-d-valine,fmoc-d-n-me-val-oh,fmoc-n-me-d-val-oh,fmoc-nalpha-methyl-d-valine,r-2-9h-fluoren-9-yl methoxy carbonyl methyl amino-3-methylbutanoic acid,2r-2-9h-fluoren-9-ylmethoxy carbonyl methyl amino-3-methylbutanoic acid,ambotzfaa1533,fmoc-d-n-me-ala-oh,fmoc-d-n-me-val-oh,n-fmoc-n-methyl-d-valine
16213159
(2R)-2-[9H-fluoren-9-ylmethoxycarbonyl(methyl)amino]-3-methylbutanoic acid
CC(C)C(C(=O)O)N(C)C(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
Formula Weight 353.42
Physical Form Solid
Percent Purity 95%
Quantity 250mg
Chemical Name or Material N-Fmoc-N-methyl-D-valine

storageNote1 : -30° to -10°C