Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Fmoc-O-benzyl-D-serine, 95%

Click to view available options
Quantity:
1g
5g
122889-11-6
C25H23NO5
417.461
MFCD00237032
DYBDGLCDMLNEMJ-HSZRJFAPSA-N
fmoc-d-ser bzl-oh,2r-3-benzyloxy-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid,r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-benzyloxy propanoic acid,ambotzfaa1380,n-fmoc-o-benzyl-d-serine,n-9h-fluoren-9-yl methoxycarbonyl-o-benzyl-d-serine,n-alpha-9-fluorenylmethyloxycarbonyl-n-im-trityl-l-histidine pentafluorphenyl ester
7019719
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-phenylmethoxypropanoic acid
C1=CC=C(C=C1)COCC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
122889-11-6
C25H23NO5
417.461
MFCD00237032
DYBDGLCDMLNEMJ-HSZRJFAPSA-N
fmoc-d-ser bzl-oh,2r-3-benzyloxy-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid,r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-benzyloxy propanoic acid,ambotzfaa1380,n-fmoc-o-benzyl-d-serine,n-9h-fluoren-9-yl methoxycarbonyl-o-benzyl-d-serine,n-alpha-9-fluorenylmethyloxycarbonyl-n-im-trityl-l-histidine pentafluorphenyl ester
7019719
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-phenylmethoxypropanoic acid
C1=CC=C(C=C1)COCC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
Formula Weight 417.46
Physical Form Solid
Percent Purity 95%
Quantity 5g
Chemical Name or Material N-Fmoc-O-benzyl-D-serine

storageNote1 : -30° to -10°C