Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Fmoc-O-benzyl-D-tyrosine, 95%

Click to view available options
Quantity:
250mg
1g
5g
138775-48-1
C31H27NO5
493.559
MFCD00235837
REHSJSKPWIOKIJ-GDLZYMKVSA-N
fmoc-d-tyr bzl-oh,fmoc-o-benzyl-d-tyr,r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-benzyloxy phenyl propanoic acid,2r-3-4-benzyloxy phenyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid,fmoc-o-benzyl-l-tyr,n-alpha-9-fluorenylmethyloxycarbonyl-o-benzyl-d-tyrosine,o-benzyl-n-9h-fluoren-9-yl methoxy carbonyl-d-tyrosine,d-tyrosine,n-9h-fluoren-9-ylmethoxy carbonyl-o-phenylmethyl
40428316
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(4-phenylmethoxyphenyl)propanoic acid
C1=CC=C(C=C1)COC2=CC=C(C=C2)CC(C(=O)O)NC(=O)OCC3C4=CC=CC=C4C5=CC=CC=C35
138775-48-1
C31H27NO5
493.559
MFCD00235837
REHSJSKPWIOKIJ-GDLZYMKVSA-N
fmoc-d-tyr bzl-oh,fmoc-o-benzyl-d-tyr,r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-benzyloxy phenyl propanoic acid,2r-3-4-benzyloxy phenyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid,fmoc-o-benzyl-l-tyr,n-alpha-9-fluorenylmethyloxycarbonyl-o-benzyl-d-tyrosine,o-benzyl-n-9h-fluoren-9-yl methoxy carbonyl-d-tyrosine,d-tyrosine,n-9h-fluoren-9-ylmethoxy carbonyl-o-phenylmethyl
40428316
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(4-phenylmethoxyphenyl)propanoic acid
C1=CC=C(C=C1)COC2=CC=C(C=C2)CC(C(=O)O)NC(=O)OCC3C4=CC=CC=C4C5=CC=CC=C35
Formula Weight 439.56
Physical Form Powder
Percent Purity 95%
Color White
Quantity 5g
Chemical Name or Material N-Fmoc-O-benzyl-D-tyrosine

storageNote1 : -30° to -10°C