Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Fmoc-O-benzyl-L-threonine, 95%

Click to view available options
Quantity:
5g
25g
117872-75-0
C26H25NO5
431.488
MFCD00077073
UCDMMWCWPVCHLL-OSPHWJPCSA-N
fmoc-thr bzl-oh,fmoc-o-benzyl-l-threonine,2s,3r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-benzyloxy butanoic acid,2s,3r-3-benzyloxy-2-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid,pubchem12952,fmoc-dl-thr bzl-oh,fmoc-thr bzl-oh tlc,n-alpha-9-fluorenylmethyloxycarbonyl-o-benzyl-l-threonine,o-benzyl-n-9h-fluoren-9-yl methoxy carbonyl-l-threonine,n-9h-fluoren-9-ylmethoxy carbonyl-l-threonine phenylmethyl ester
16213170
(2S,3R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-phenylmethoxybutanoic acid
CC(C(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13)OCC4=CC=CC=C4
117872-75-0
C26H25NO5
431.488
MFCD00077073
UCDMMWCWPVCHLL-OSPHWJPCSA-N
fmoc-thr bzl-oh,fmoc-o-benzyl-l-threonine,2s,3r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-benzyloxy butanoic acid,2s,3r-3-benzyloxy-2-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid,pubchem12952,fmoc-dl-thr bzl-oh,fmoc-thr bzl-oh tlc,n-alpha-9-fluorenylmethyloxycarbonyl-o-benzyl-l-threonine,o-benzyl-n-9h-fluoren-9-yl methoxy carbonyl-l-threonine,n-9h-fluoren-9-ylmethoxy carbonyl-l-threonine phenylmethyl ester
16213170
(2S,3R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-phenylmethoxybutanoic acid
CC(C(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13)OCC4=CC=CC=C4
Beilstein 6353662
Formula Weight 431.49
Physical Form Powder
Percent Purity 95%
Color White
Quantity 5g
Chemical Name or Material N-Fmoc-O-benzyl-L-threonine

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold