Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Fmoc-O-benzylphospho-L-tyrosine, 95%

Click to view available options
Quantity:
250mg
1g
191348-16-0
C31H28NO8P
573.538
MFCD00797871
WNFKGEXQEWKECX-LJAQVGFWSA-N
fmoc-tyr hpo3bzl-oh,fmoc-tyr po obzl oh-oh,fmoc-l-tyr po obzl oh-oh,n-fmoc-o-benzyl-l-phosphotyrosine,2s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-benzyloxy hydroxy phosphoryl oxy phenyl propanoic acid,ambotzfaa1432,fmoc-tyr ho3bzl-oh,fmoc-tyr po obz oh-oh,fmoc-tyr po obzl-oh,fmoc-tyrosine po obzl oh-oh
46735206
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[4-[hydroxy(phenylmethoxy)phosphoryl]oxyphenyl]propanoic acid
C1=CC=C(C=C1)COP(=O)(O)OC2=CC=C(C=C2)CC(C(=O)O)NC(=O)OCC3C4=CC=CC=C4C5=CC=CC=C35
191348-16-0
C31H28NO8P
573.538
MFCD00797871
WNFKGEXQEWKECX-LJAQVGFWSA-N
fmoc-tyr hpo3bzl-oh,fmoc-tyr po obzl oh-oh,fmoc-l-tyr po obzl oh-oh,n-fmoc-o-benzyl-l-phosphotyrosine,2s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-benzyloxy hydroxy phosphoryl oxy phenyl propanoic acid,ambotzfaa1432,fmoc-tyr ho3bzl-oh,fmoc-tyr po obz oh-oh,fmoc-tyr po obzl-oh,fmoc-tyrosine po obzl oh-oh
46735206
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[4-[hydroxy(phenylmethoxy)phosphoryl]oxyphenyl]propanoic acid
C1=CC=C(C=C1)COP(=O)(O)OC2=CC=C(C=C2)CC(C(=O)O)NC(=O)OCC3C4=CC=CC=C4C5=CC=CC=C35
Applications
N-Fmoc-O-benzylphospho-L-tyrosine is used as pharmaceutical intermediates.

Solubility
Soluble in 0.1g in 2mL DMF.

Notes
Protect from heat. Store away from oxidizing agents.
Formula Weight 573.54
Physical Form Powder
Percent Purity 95%
Color White
Quantity 1g
Chemical Name or Material N-Fmoc-O-benzylphospho-L-tyrosine

storageNote1 : -30° to -10°C