Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Fmoc-O-tert-butyl-D-allo-threonine, 98%

Click to view available options
Quantity:
250mg
1g
170643-02-4
C23H27NO5
397.471
MFCD00077074
LZOLWEQBVPVDPR-JLTOFOAXSA-N
fmoc-d-allo-thr tbu-oh,2r,3r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-tert-butoxy butanoic acid,fmoc-d-allo-threonine tbu-oh,nalpha-9-fluorenylmethoxycarbonyl-o-t-butyl-d-allo-threonine,n-fmoc-o-tert-butyl-d-allothreonine,n-9h-fluoren-9-yl methoxy carbonyl-o-tert-butyl-d-allothreonine,o-tert-butyl-n-9h-fluoren-9-ylmethoxy carbonyl-d-allothreonine,o-tert-butyl-n-9h-fluoren-9-yl methoxy carbonyl-d-allothreonine,2r,3r-3-tert-butoxy-2-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid,d-allothreonine,o-1,1-dimethylethyl-n-9h-fluoren-9-ylmethoxy carbonyl
10949330
(2R,3R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[(2-methylpropan-2-yl)oxy]butanoic acid
CC(C(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13)OC(C)(C)C
170643-02-4
C23H27NO5
397.471
MFCD00077074
LZOLWEQBVPVDPR-JLTOFOAXSA-N
fmoc-d-allo-thr tbu-oh,2r,3r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-tert-butoxy butanoic acid,fmoc-d-allo-threonine tbu-oh,nalpha-9-fluorenylmethoxycarbonyl-o-t-butyl-d-allo-threonine,n-fmoc-o-tert-butyl-d-allothreonine,n-9h-fluoren-9-yl methoxy carbonyl-o-tert-butyl-d-allothreonine,o-tert-butyl-n-9h-fluoren-9-ylmethoxy carbonyl-d-allothreonine,o-tert-butyl-n-9h-fluoren-9-yl methoxy carbonyl-d-allothreonine,2r,3r-3-tert-butoxy-2-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid,d-allothreonine,o-1,1-dimethylethyl-n-9h-fluoren-9-ylmethoxy carbonyl
10949330
(2R,3R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[(2-methylpropan-2-yl)oxy]butanoic acid
CC(C(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13)OC(C)(C)C
Formula Weight 397.47
Physical Form Powder
Percent Purity 98%
Color White
Quantity 250mg
Chemical Name or Material N-Fmoc-O-tert-butyl-D-allo-threonine

storageNote1 : -30° to -10°C