Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Fmoc-O-tert-butyl-D-threonine, 95%

Click to view available options
Quantity:
1g
5g
138797-71-4
C23H27NO5
397.471
MFCD00080292
LZOLWEQBVPVDPR-VBKZILBWSA-N
fmoc-d-thr tbu-oh,fmoc-o-tert-butyl-d-threonine,fmoc-d-thr but-oh,fmoc-thr but-oh,2r,3s-3-tert-butoxy-2-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid,2s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-tert-butoxy butanoic acid,fmoc-o-tert.butyl-d-threonine,ambotzfaa1509,pubchem10044,n-fmoc-o-tert-butyl-d-threonine
7021418
(2R,3S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[(2-methylpropan-2-yl)oxy]butanoic acid
CC(C(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13)OC(C)(C)C
138797-71-4
C23H27NO5
397.471
MFCD00080292
LZOLWEQBVPVDPR-VBKZILBWSA-N
fmoc-d-thr tbu-oh,fmoc-o-tert-butyl-d-threonine,fmoc-d-thr but-oh,fmoc-thr but-oh,2r,3s-3-tert-butoxy-2-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid,2s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-tert-butoxy butanoic acid,fmoc-o-tert.butyl-d-threonine,ambotzfaa1509,pubchem10044,n-fmoc-o-tert-butyl-d-threonine
7021418
(2R,3S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[(2-methylpropan-2-yl)oxy]butanoic acid
CC(C(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13)OC(C)(C)C
Beilstein 8796152
Formula Weight 397.47
Physical Form Powder
Percent Purity 95%
Color Yellow
Melting Point ~130°C
Quantity 5g
Chemical Name or Material N-Fmoc-O-tert-butyl-D-threonine

storageNote1 : 2° to 8°C