Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Fmoc-O-tert-butyl-L-allo-threonine, 97%

Click to view available options
Quantity:
250mg
1g
5g
201481-37-0
C23H27NO5
397.471
MFCD00237371
LZOLWEQBVPVDPR-XOBRGWDASA-N
fmoc-allo-thr tbu-oh,fmoc-l-allo-thr tbu-oh,2s,3s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-tert-butoxy butanoic acid,fmoc-allo-thr tbu,ambotzfaa1655,o-tert-butyl-n-fmoc-l-allothreonine,n-alpha-9-fluorenylmethyloxycarbonyl-o-t-butyl-allo-l-threonine,2s,3s-2-9h-fluoren-9-ylmethoxycarbonylamino-3-2-methylpropan-2-yl oxy butanoic acid,2s,3s-3-tert-butoxy-2-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid
2724634
(2S,3S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[(2-methylpropan-2-yl)oxy]butanoic acid
CC(C(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13)OC(C)(C)C
201481-37-0
C23H27NO5
397.471
MFCD00237371
LZOLWEQBVPVDPR-XOBRGWDASA-N
fmoc-allo-thr tbu-oh,fmoc-l-allo-thr tbu-oh,2s,3s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-tert-butoxy butanoic acid,fmoc-allo-thr tbu,ambotzfaa1655,o-tert-butyl-n-fmoc-l-allothreonine,n-alpha-9-fluorenylmethyloxycarbonyl-o-t-butyl-allo-l-threonine,2s,3s-2-9h-fluoren-9-ylmethoxycarbonylamino-3-2-methylpropan-2-yl oxy butanoic acid,2s,3s-3-tert-butoxy-2-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid
2724634
(2S,3S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[(2-methylpropan-2-yl)oxy]butanoic acid
CC(C(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13)OC(C)(C)C
Formula Weight 397.47
Physical Form Solid
Percent Purity 97%
Color White
Quantity 5g
Chemical Name or Material N-Fmoc-O-tert-butyl-L-allo-threonine

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C