Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Fmoc-O-tert-butyl-L-tyrosine pentafluorophenyl ester, 96%

Click to view available options
Quantity:
1g
5g
86060-93-7
C34H28F5NO5
625.592
MFCD00077077
ADOSTZDWXCEVJP-VWLOTQADSA-N
fmoc-tyr tbu-opfp,fmoc-o-t-butyl-l-tyrosine pentafluorophenyl ester,fmoc-o-tert-butyl-l-tyrosine pentafluorophenyl ester,l-tyrosine,o-1,1-dimethylethyl-n-9h-fluoren-9-ylmethoxy carbonyl-,2,3,4,5,6-pentafluorophenyl ester,2,3,4,5,6-pentafluorophenyl 2s-3-4-tert-butoxy phenyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoate,fmoc-tyr tbu-opfp hplc,n-fmoc-o-tert-butyl-l-tyrosine pentafluorophenyl ester,n-9h-fluoren-9-ylmethoxycarbonyl-o-tert-butyl-l-tyrosine pentafluorophenyl ester,pentafluorophenyl o-tert-butyl-n-9h-fluoren-9-ylmethoxy carbonyl-l-tyrosinate
10886688
(2,3,4,5,6-pentafluorophenyl) (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[4-[(2-methylpropan-2-yl)oxy]phenyl]propanoate
CC(C)(C)OC1=CC=C(C=C1)CC(C(=O)OC2=C(C(=C(C(=C2F)F)F)F)F)NC(=O)OCC3C4=CC=CC=C4C5=CC=CC=C35
86060-93-7
C34H28F5NO5
625.592
MFCD00077077
ADOSTZDWXCEVJP-VWLOTQADSA-N
fmoc-tyr tbu-opfp,fmoc-o-t-butyl-l-tyrosine pentafluorophenyl ester,fmoc-o-tert-butyl-l-tyrosine pentafluorophenyl ester,l-tyrosine,o-1,1-dimethylethyl-n-9h-fluoren-9-ylmethoxy carbonyl-,2,3,4,5,6-pentafluorophenyl ester,2,3,4,5,6-pentafluorophenyl 2s-3-4-tert-butoxy phenyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoate,fmoc-tyr tbu-opfp hplc,n-fmoc-o-tert-butyl-l-tyrosine pentafluorophenyl ester,n-9h-fluoren-9-ylmethoxycarbonyl-o-tert-butyl-l-tyrosine pentafluorophenyl ester,pentafluorophenyl o-tert-butyl-n-9h-fluoren-9-ylmethoxy carbonyl-l-tyrosinate
10886688
(2,3,4,5,6-pentafluorophenyl) (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[4-[(2-methylpropan-2-yl)oxy]phenyl]propanoate
CC(C)(C)OC1=CC=C(C=C1)CC(C(=O)OC2=C(C(=C(C(=C2F)F)F)F)F)NC(=O)OCC3C4=CC=CC=C4C5=CC=CC=C35
Beilstein 4223194
Formula Weight 625.59
Physical Form Powder
Percent Purity 96%
Color White
Quantity 1g
Chemical Name or Material N-Fmoc-O-tert-butyl-L-tyrosine pentafluorophenyl ester

storageNote1 : 2° to 8°C