Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Fmoc-O-tert-butyl-N-methyl-L-serine, 97%

Click to view available options
Quantity:
250mg
1g
5g
197632-77-2
C23H27NO5
397.471
MFCD02094430
PQSAXALAXPNFMG-FQEVSTJZSA-N
fmoc-n-me-ser tbu-oh,n-fmoc-n-methyl-o-tert-butyl-l-serine,fmoc-n-methyl-o-t-butyl-l-serine,fmoc-n-methyl-o-tert-butyl-l-serine,n-alpha-9-fluorenylmethyloxycarbonyl-n-alpha-methyl-o-t-butyl-l-serine,2s-3-tert-butoxy-2-9h-fluoren-9-ylmethoxy carbonyl methyl amino propanoic acid,s-2-9h-fluoren-9-yl methoxy carbonyl methyl amino-3-tert-butoxy propanoic acid,fmoc-meser tbu-oh,pubchem19046,fmoc-n-me-serine tbu-oh
7010370
(2S)-2-[9H-fluoren-9-ylmethoxycarbonyl(methyl)amino]-3-[(2-methylpropan-2-yl)oxy]propanoic acid
CC(C)(C)OCC(C(=O)O)N(C)C(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
197632-77-2
C23H27NO5
397.471
MFCD02094430
PQSAXALAXPNFMG-FQEVSTJZSA-N
fmoc-n-me-ser tbu-oh,n-fmoc-n-methyl-o-tert-butyl-l-serine,fmoc-n-methyl-o-t-butyl-l-serine,fmoc-n-methyl-o-tert-butyl-l-serine,n-alpha-9-fluorenylmethyloxycarbonyl-n-alpha-methyl-o-t-butyl-l-serine,2s-3-tert-butoxy-2-9h-fluoren-9-ylmethoxy carbonyl methyl amino propanoic acid,s-2-9h-fluoren-9-yl methoxy carbonyl methyl amino-3-tert-butoxy propanoic acid,fmoc-meser tbu-oh,pubchem19046,fmoc-n-me-serine tbu-oh
7010370
(2S)-2-[9H-fluoren-9-ylmethoxycarbonyl(methyl)amino]-3-[(2-methylpropan-2-yl)oxy]propanoic acid
CC(C)(C)OCC(C(=O)O)N(C)C(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
Formula Weight 397.46
Physical Form Crystalline Solid
Percent Purity 97%
Color White
Melting Point 213°C to 218°C
Quantity 1g
Chemical Name or Material N-Fmoc-O-tert-butyl-N-methyl-L-serine

TSCA : No

Recommended Storage : Ambient temperatures