Alfa Aesar™ N-Fmoc-S-trityl-L-cysteine, 95%

Manufacturer: Alfa Aesar™ H2736303

Quantity 1g

 View more versions of this product

Catalog No. AAH2736303

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 103213-32-7
    Molecular Formula C37H31NO4S
    Formula Weight 585.71
    MDL Number MFCD00038538
    Synonym N-(9-Fluorenylmethoxycarbonyl)-S-trityl-L-cysteine; Fmoc-Cys(Trt)-OH
    Chemical Name or Material N-Fmoc-S-trityl-L-cysteine

    Specifications

    Formula Weight 585.71
    Melting Point 171° to 173°C
    MDL Number MFCD00038538
    Quantity 1g
    Optical Rotation +17° (c=1 in THF)
    Molecular Formula C37H31NO4S
    CAS 103213-32-7
    Synonym N-(9-Fluorenylmethoxycarbonyl)-S-trityl-L-cysteine; Fmoc-Cys(Trt)-OH
    Recommended Storage 2° to 8°C
    Chemical Name or Material N-Fmoc-S-trityl-L-cysteine
    Assay 95%

    Description

    Applications
    It is potentially useful for proteomics studies, and solid phase peptide synthesis techniques. Cysteine is versatile amino acid involved with many biological processes, including the formation of disulfide bonds - a critical component of protein structure. This compound could be useful as an unusual amino acid analog to aid in the deconvolution of protein structure and function.

    Solubility
    Insoluble in water. Soluble in most organic solvents.

    Notes
    Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

    2° to 8°C

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit