Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Glycyl-L-tyrosine, 98%

Click to view available options
Quantity:
5g
25g
100g
658-79-7
C11H14N2O4
238.243
MFCD00008126
XBGGUPMXALFZOT-VIFPVBQESA-N
glycyl-l-tyrosine,gly-tyr,n-glycyl-l-tyrosine,h-gly-tyr-oh,glycyltyrosine,l-tyrosine, glycyl,s-2-2-aminoacetamido-3-4-hydroxyphenyl propanoic acid,chembl53400,2s-2-2-aminoacetamido-3-4-hydroxyphenyl propanoic acid,2s-2-2-aminoacetyl amino-3-4-hydroxyphenyl propanoic acid
92829
CHEBI:73517
(2S)-2-[(2-aminoacetyl)amino]-3-(4-hydroxyphenyl)propanoic acid
C1=CC(=CC=C1CC(C(=O)O)NC(=O)CN)O
658-79-7
C11H14N2O4
238.243
MFCD00008126
XBGGUPMXALFZOT-VIFPVBQESA-N
glycyl-l-tyrosine,gly-tyr,n-glycyl-l-tyrosine,h-gly-tyr-oh,glycyltyrosine,l-tyrosine, glycyl,s-2-2-aminoacetamido-3-4-hydroxyphenyl propanoic acid,chembl53400,2s-2-2-aminoacetamido-3-4-hydroxyphenyl propanoic acid,2s-2-2-aminoacetyl amino-3-4-hydroxyphenyl propanoic acid
92829
CHEBI:73517
(2S)-2-[(2-aminoacetyl)amino]-3-(4-hydroxyphenyl)propanoic acid
C1=CC(=CC=C1CC(C(=O)O)NC(=O)CN)O
Beilstein 2700715
Formula Weight 238.24
Physical Form Solid
Percent Purity 98%
Color White to Yellow
Quantity 100g
Chemical Name or Material N-Glycyl-L-tyrosine

storageNote1 : -30° to -10°C