Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Heneicosane, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
10g
50g
Packaging:
Glass bottle
629-94-7
C21H44
296.58
MFCD00009346
FNAZRRHPUDJQCJ-UHFFFAOYSA-N
heneicosane,n-heneicosane,unii-i93s5u5dmp,eicosane, methyl,i93s5u5dmp,ch3-ch2 19-ch3,henicosan,acmc-1b9xj
12403
CHEBI:32931
henicosane
CCCCCCCCCCCCCCCCCCCCC
629-94-7
C21H44
296.58
MFCD00009346
FNAZRRHPUDJQCJ-UHFFFAOYSA-N
heneicosane,n-heneicosane,unii-i93s5u5dmp,eicosane, methyl,i93s5u5dmp,ch3-ch2 19-ch3,henicosan,acmc-1b9xj
12403
CHEBI:32931
henicosane
CCCCCCCCCCCCCCCCCCCCC
Packaging Glass bottle
Boiling Point 100°C (2.0mmHg)
Flash Point >110°C
Color White
Quantity 1g
Melting Point 39°C to 43°C
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula CH3(CH2)19CH3
Solubility Information Solubility in water: insoluble. Other solubilities: slightly soluble in ethanol,,soluble in petroleum ether
Formula Weight 296.58
Physical Form Crystalline Chunks, Crystals, or Flakes
Percent Purity 98%
Chemical Name or Material n-Heneicosane
Show More Show Less

EINECSNumber : 211-118-9