n-Heptadecane, 99%, Alfa Aesar™

Manufacturer: Alfa Aesar 3155114

Quantity 25g

 View more versions of this product

Catalog No. AA3155114

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 629-78-7
    Molecular Formula CH3(CH2)15CH3
    Formula Weight 240.48
    MDL Number MFCD00009002
    Chemical Name or Material n-Heptadecane

    Specifications

    Physical Form Low Melting Solid
    Formula Weight 240.48
    Quantity 25g
    TSCA TSCA
    MDL Number MFCD00009002
    Health Hazard 3 GHS P Statement
    P301+P310-P331-P405-P501a;
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    RTECS Number MI3550000
    Molecular Formula CH3(CH2)15CH3
    Refractive Index 1.4369
    CAS 629-78-7
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H304;
    May be fatal if swallowed and enters airways.
    Density 0.778
    Boiling Point 302°C
    Melting Point 22°C
    Chemical Name or Material n-Heptadecane
    Assay 99%

    GHS H Statement
    H304;
    May be fatal if swallowed and enters airways.

    GHS P Statement
    P301+P310-P331-P405-P501a;
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit