Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Heptane, 99.5%, for analysis, ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
5L
25L
142-82-5
C7H16
100.205
MFCD00009544
IMNFDUFMRHMDMM-UHFFFAOYSA-N
n-heptane,dipropylmethane,heptan,heptyl hydride,dipropyl methane,gettysolve-c,skellysolve c,heptanen,eptani,heptan polish
8900
CHEBI:43098
heptane
CCCCCCC
142-82-5
C7H16
100.205
MFCD00009544
IMNFDUFMRHMDMM-UHFFFAOYSA-N
n-heptane,dipropylmethane,heptan,heptyl hydride,dipropyl methane,gettysolve-c,skellysolve c,heptanen,eptani,heptan polish
8900
CHEBI:43098
heptane
CCCCCCC
For analysis
CAS Min % 99.4
CAS Max % 100.0
Acidity 0.0001 meq/g max.
Assay Percent Range 99.5%
Linear Formula CH3(CH2)5CH3
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in alcohol, ether and chloroform
Bromine Number 0.5 max.
Calcium (Ca) 0.5ppm max.
Formula Weight 100.2
Physical Form Liquid
Percent Purity 99.5%
Grade Analysis
Copper (Cu) 0.02ppm max.
Density 0.6830g/mL
Iron (Fe) 0.1ppm max.
Lead (Pb) 0.02ppm max.
Magnesium (Mg) 0.05ppm max.
Packaging Plastic drum
Potassium (K) 0.1ppm max.
Refractive Index 1.3850 to 1.3890
Residue after Evaporation 0.001% max.
Silicon (Si) 0.05ppm max.
Solubility Solubility in water: practically insoluble. Other solubilities: soluble in alcohol, ether and chloroform
Sodium (Na) 0.5ppm max.
Specific Gravity 0.683
Substances Darkened by H2SO4 10 APHA max.
Total Phosphorus 0.1ppm max.
Trace Analysis 0.2ppm max.
Total Sulfur 0.001% max.
Water 0.02% max. (K.F.)
Flash Point −4°C
Viscosity 0.4 mPa.s (20°C)
Boiling Point 98.0°C
Color Undesignated
Melting Point -91.0°C
Quantity 5L
Chemical Name or Material n-Heptane
Show More Show Less

GHS H Statement
Causes skin irritation. May cause drowsiness or dizziness. May be fatal if swallowed and enters airways. Highly flammable liquid and vapour. Very toxic to aquatic life with long lasting effects.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. Avoid release to the environment. IF ON SKIN: Wash with plenty of soap and water. Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

Danger