Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Heptane, 99%, for biochemistry, AcroSeal™, Acros Organics

Click to view available options
Quantity:
1L
2.5L
142-82-5
C7H16
100.2
IMNFDUFMRHMDMM-UHFFFAOYSA-N
n-heptane,dipropylmethane,heptan,heptyl hydride,dipropyl methane,gettysolve-c,skellysolve c,heptanen,eptani,heptan polish
8900
CHEBI:43098
heptane
CCCCCCC
142-82-5
C7H16
100.2
IMNFDUFMRHMDMM-UHFFFAOYSA-N
n-heptane,dipropylmethane,heptan,heptyl hydride,dipropyl methane,gettysolve-c,skellysolve c,heptanen,eptani,heptan polish
8900
CHEBI:43098
heptane
CCCCCCC
Viscosity 0.4 mPa.s (20°C)
Flash Point −4°C
Quantity 2.5L
Refractive Index 1.3850 to 1.3890
Boiling Point 98.0°C
Specific Gravity 0.683
Density 0.6830g/mL
Melting Point -91.0°C
Assay Percent Range 98.5% min. (GC)
Linear Formula CH3(CH2)5CH3
Merck Index 15,4694
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in alcohol, ether and chloroform
Formula Weight 100.2
Percent Purity 99%
Grade Biochemistry
Chemical Name or Material n-Heptane
Show More Show Less

GHS H Statement
Causes skin irritation.
May cause drowsiness or dizziness.
May be fatal if swallowed and enters airways.
Highly flammable liquid and vapour.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap

GHS Signal Word: Danger

EINECSNumber : 205-563-8