Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Heptane, 99+%, extra pure, ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
5L
25L
142-82-5
C7H16
100.205
IMNFDUFMRHMDMM-UHFFFAOYSA-N
n-heptane,dipropylmethane,heptan,heptyl hydride,dipropyl methane,gettysolve-c,skellysolve c,heptanen,eptani,heptan polish
8900
CHEBI:43098
heptane
CCCCCCC
142-82-5
C7H16
100.205
IMNFDUFMRHMDMM-UHFFFAOYSA-N
n-heptane,dipropylmethane,heptan,heptyl hydride,dipropyl methane,gettysolve-c,skellysolve c,heptanen,eptani,heptan polish
8900
CHEBI:43098
heptane
CCCCCCC
Acidity 0.003 meq/g max.
Assay Percent Range 99+%
Linear Formula CH3(CH2)5CH3
Merck Index 15, 4694
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in alcohol, ether and chloroform
Calcium (Ca) 2ppm max.
Formula Weight 100.2
Physical Form Liquid
Percent Purity ≥99%
Grade Extra Pure
Copper (Cu) 1ppm max.
Density 0.6830g/mL
Infrared Spectrum Authentic
Iron (Fe) 5ppm max.
Lead (Pb) 1ppm max.
Magnesium (Mg) 1ppm max.
Packaging Plastic drum
Potassium (K) 1ppm max.
Refractive Index 1.3850 to 1.3890
Residue after Evaporation 20ppm max.
Silicon (Si) 1ppm max.
Solubility Solubility in water: practically insoluble. Other solubilities: soluble in alcohol, ether and chloroform
Sodium (Na) 2ppm max.
Specific Gravity 0.683
Substances Darkened by H2SO4 20 APHA max.
Total Phosphorus 1ppm max.
Total Sulfur 50ppm max.
Water 0.05% max.
Zinc (Zn) 2ppm max.
Flash Point −4°C
Viscosity 0.4 mPa.s (20°C)
Boiling Point 98.0°C
Color Colorless to Yellow
Melting Point -91.0°C
Quantity 5L
Chemical Name or Material n-Heptane
Show More Show Less

GHS H Statement
Causes skin irritation. May cause drowsiness or dizziness. May be fatal if swallowed and enters airways. Highly flammable liquid and vapour. Very toxic to aquatic life with long lasting effects.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. Avoid release to the environment. IF ON SKIN: Wash with plenty of soap and water. Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

Danger