Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Heptane, 99+%, pure, ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
5L
25L
142-82-5
C7H16
100.205
MFCD00009544
IMNFDUFMRHMDMM-UHFFFAOYSA-N
n-heptane,dipropylmethane,heptan,heptyl hydride,dipropyl methane,gettysolve-c,skellysolve c,heptanen,eptani,heptan polish
8900
CHEBI:43098
heptane
CCCCCCC
142-82-5
C7H16
100.205
MFCD00009544
IMNFDUFMRHMDMM-UHFFFAOYSA-N
n-heptane,dipropylmethane,heptan,heptyl hydride,dipropyl methane,gettysolve-c,skellysolve c,heptanen,eptani,heptan polish
8900
CHEBI:43098
heptane
CCCCCCC
CAS Min % 99.0
CAS Max % 100.0
Additional Information Vapor Pressure: 48mbar at 20°C
Assay Percent Range 99+%
Linear Formula CH3(CH2)5CH3
Beilstein 01, 154
Merck Index 15, 4694
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in alcohol, ether and chloroform
Formula Weight 100.2
Physical Form Liquid
Percent Purity ≥99%
Grade Pure
Density 0.6830g/mL
Infrared Spectrum Authentic
Packaging Glass Bottle
Refractive Index 1.3850 to 1.3890
Solubility Solubility in water: practically insoluble. Other solubilities: soluble in alcohol, ether and chloroform
Specific Gravity 0.683
Flash Point −4°C
Viscosity 0.4 mPa.s (20°C)
Boiling Point 98.0°C
Color Colorless
Melting Point -91.0°C
Quantity 1L
Chemical Name or Material n-Heptane
Show More Show Less

GHS P Statement Causes skin irritation. May cause drowsiness or dizziness. May be fatal if swallowed and enters airways. Highly flammable liquid and vapour. Very toxic to aquatic life with long lasting effects.

GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. Avoid release to the environment. IF ON SKIN: Wash with plen

Danger