Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Heptane, 99+%, for spectroscopy, ACROS Organics™

Click to view available options
Quantity:
500mL
2.5L
Packaging:
EcoSafPak™ Glass Bottle
Glass bottle
142-82-5
C7H16
100.21
MFCD00009544
IMNFDUFMRHMDMM-UHFFFAOYSA-N
n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
8900
CHEBI:43098
heptane
CCCCCCC
142-82-5
C7H16
100.21
MFCD00009544
IMNFDUFMRHMDMM-UHFFFAOYSA-N
n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
8900
CHEBI:43098
heptane
CCCCCCC
For UV/VIS spectroscopy
Absorbance 0.01 max. at 240nm, 0.01 max. at 250nm, 0.01 max. at 260nm, 0.01 max. at 400nm, 0.05 max. at 230nm, 0.15 max. at 220nm, 0.4 max. at 210nm, 1 max. at 197nm, 1 max. at 200nm
CAS Min % 99.0
CAS Max % 100.0
Additional Information Vapor Pressure: 48mbar at 20°C
Assay Percent Range 99+%
Linear Formula CH3(CH2)5CH3
Beilstein 01, 154
Merck Index 15, 4694
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in alcohol, ether and chloroform
Formula Weight 100.2
Physical Form Liquid
Percent Purity ≥99%
Grade Spectroscopy
Density 0.6830g/mL
Packaging EcoSafPak™ Glass Bottle
Refractive Index 1.3850 to 1.3890
Residue after Evaporation 0.0005 % max.
Specific Gravity 0.683
Water 0.03% Max.
Flash Point −4°C
Viscosity 0.4 mPa.s (20°C)
Boiling Point 98.0°C
Color Undesignated
Melting Point -91.0°C
Quantity 500mL
Chemical Name or Material n-Heptane
Show More Show Less

GHS P Statement Causes skin irritation. May cause drowsiness or dizziness. May be fatal if swallowed and enters airways. Highly flammable liquid and vapour. Very toxic to aquatic life with long lasting effects.

GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. Avoid release to the environment. IF ON SKIN: Wash with plen

Danger

EINECSNumber : 205-563-8

RTECSNumber : MI7700000

TSCA : TSCA