Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Heptane, BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
500mL
4L
Packaging:
Narrow Mouth Amber Glass Bottle
Narrow Mouth PVC Coated Amber Bottle
142-82-5
C7H16
100.205
IMNFDUFMRHMDMM-UHFFFAOYSA-N
n-heptane,dipropylmethane,heptan,heptyl hydride,dipropyl methane,gettysolve-c,skellysolve c,heptanen,eptani,heptan polish
8900
CHEBI:43098
heptane
CCCCCCC
142-82-5
C7H16
100.205
IMNFDUFMRHMDMM-UHFFFAOYSA-N
n-heptane,dipropylmethane,heptan,heptyl hydride,dipropyl methane,gettysolve-c,skellysolve c,heptanen,eptani,heptan polish
8900
CHEBI:43098
heptane
CCCCCCC

High quality chemicals for laboratory and specialized industrial use

Formula Weight 100.2
Grade BAKER ANALYZED™ Reagent
Packaging Narrow Mouth PVC Coated Amber Bottle
Specific Gravity 1L = 0.68kg
Flash Point −4.00°C
Quantity 4L
Chemical Name or Material n-Heptane