Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Heptane, BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
4L
500mL
Packaging:
Narrow Mouth Amber Glass Bottle
Narrow Mouth PVC Coated Amber Bottle

Chemical Identifiers

142-82-5
C7H16
100.205
IMNFDUFMRHMDMM-UHFFFAOYSA-N
8900
CHEBI:43098

High quality chemicals for laboratory and specialized industrial use

CAS 142-82-5
Molecular Formula C7H16
InChI Key IMNFDUFMRHMDMM-UHFFFAOYSA-N
SMILES CCCCCCC
IUPAC Name heptane
Molecular Weight (g/mol) 100.205
PubChem CID 8900
ChEBI CHEBI:43098
Formula Weight 100.2
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 0.68kg
Flash Point −4.00°C
Quantity 4L
Chemical Name or Material n-Heptane
Grade BAKER ANALYZED™ Reagent
Show More Show Less