Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Heptane, Environmental Grade, 96+%, Alfa Aesar™

Click to view available options
Quantity:
1L
4L
142-82-5
C7H16
100.21
MFCD00009544
IMNFDUFMRHMDMM-UHFFFAOYSA-N
n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
8900
CHEBI:43098
heptane
CCCCCCC
142-82-5
C7H16
100.21
MFCD00009544
IMNFDUFMRHMDMM-UHFFFAOYSA-N
n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
8900
CHEBI:43098
heptane
CCCCCCC

For Pesticide Residue, Environmental, and GC Analysis

Submicron filtered, halogenated residue: 10ng/L (GC-ECD)
Assay Percent Range >96%
Appearance Colorless liquid
Beilstein 1730763
Solubility Information Miscible in alcohol,chloroform,and ether. Insoluble in water.
Formula Weight 100.21
Physical Form Liquid
Grade Environmental
Density 0.684 g/cm3 at 20°C
Ignition Point 215°C (419°F)
Odor Petroleum-like
Refractive Index 1.3877
Flash Point -4°C (24°F)
Boiling Point 97°C to 99°C
Color Colorless
Melting Point -90°C
Quantity 1L
Chemical Name or Material n-Heptane
Show More Show Less

Keep container in a well ventilated place. Keep away from sources of ignition - No Smoking. Do not empty into drains. Take precautionary measures against static discharges. This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets If swallowed, do not induce vomiting: seek medical advice immediately & show this container or label.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P331-P332+P313-P363-P370+P378q-P501c

H225-H304-H315-H319-H336-H373

DOTInformation : DOT Class: 3, Packaging Group: II

EINECSNumber : 205-563-8

RTECSNumber : MI7700000

TSCA : Yes

Recommended Storage : Ambient temperatures