n-Heptane, Environmental Grade, 96+%, Alfa Aesar™

For Pesticide Residue, Environmental, and GC Analysis

Manufacturer: Alfa Aesar 40978K7

Quantity 4L

 View more versions of this product

Catalog No. AA40978K7

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 142-82-5
    Physical Form Liquid
    Assay Percent Range >96%
    Molecular Formula CH3(CH2)5CH3
    Formula Weight 100.21
    MDL Number MFCD00009544
    Chemical Name or Material n-Heptane
    Grade Environmental Grade

    Specifications

    Physical Form Liquid
    Color Clear colorless
    Formula Weight 100.21
    Quantity 4L
    DOT Information DOT Class: 3, Packaging Group: II
    Health Hazard 2 Keep container in a well ventilated place. Keep away from sources of ignition - No Smoking. Do not empty into drains. Take precautionary measures against static discharges. This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets If swallowed, do not induce vomiting: seek medical advice immediately & show this container or label.
    Density 0.684g/cm3 at 20°C
    EINECS Number 205-563-8
    Ignition Point 215°C (419°F)
    Odor Petroleum-like
    Appearance Colorless liquid
    Assay Percent Range >96%
    Molecular Formula CH3(CH2)5CH3
    MDL Number MFCD00009544
    CAS 142-82-5
    Health Hazard 1 H225-H304-H400-H410-H315-H336: Highly flammable liquid and vapour. May be fatal if swallowed and enters airways. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. Causes skin irritation. May cause drowsiness or dizziness.
    Chemical Name or Material n-Heptane
    Grade Environmental Grade

    Keep container in a well ventilated place. Keep away from sources of ignition - No Smoking. Do not empty into drains. Take precautionary measures against static discharges. This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets If swallowed, do not induce vomiting: seek medical advice immediately & show this container or label.

    H225-H304-H400-H410-H315-H336: Highly flammable liquid and vapour. May be fatal if swallowed and enters airways. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. Causes skin irritation. May cause drowsiness or dizziness.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit