Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Heptane, for HPLC, (approx. 99% n-Heptane), ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
142-82-5
C7H16
100.205
MFCD00009544
IMNFDUFMRHMDMM-UHFFFAOYSA-N
n-heptane,dipropylmethane,heptan,heptyl hydride,dipropyl methane,gettysolve-c,skellysolve c,heptanen,eptani,heptan polish
8900
CHEBI:43098
heptane
CCCCCCC
142-82-5
C7H16
100.205
MFCD00009544
IMNFDUFMRHMDMM-UHFFFAOYSA-N
n-heptane,dipropylmethane,heptan,heptyl hydride,dipropyl methane,gettysolve-c,skellysolve c,heptanen,eptani,heptan polish
8900
CHEBI:43098
heptane
CCCCCCC
For HPLC
Absorbance (1 cm cell vs HPLC−grade water),(1 cm cell vs HPLC−grade water) at 0nm,0.005 max. at 250nm,0.01 max. at 240nm,0.02 max. at 230nm,0.05 max. at 220nm,0.1 max. at 210nm,0.25 max. at 200nm
Absorbance Conditions (1 cm cell vs HPLC-grade water)
Acidity 0.00015 meq/g max.
Linear Formula CH3(CH2)5CH3
Beilstein 01, 154
Merck Index 15, 4694
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in alcohol, ether and chloroform
Formula Weight 100.2
Physical Form Liquid
Percent Purity ≥99% (GC)
Grade HPLC
Density 0.6830g/mL
Filtered to 0.2 micrometer Yes
Refractive Index 1.3850 to 1.3890
Residue after Evaporation 5ppm max.
Solubility Solubility in water: practically insoluble. Other solubilities: soluble in alcohol, ether and chloroform
Specific Gravity 0.683
Water 0.01% Max.
Flash Point −4°C
Viscosity 0.4 mPa.s (20°C)
Boiling Point 98.0°C
Color Colorless
Melting Point -91.0°C
Quantity 2.5L
Chemical Name or Material n-Heptane
Show More Show Less

GHS H Statement
Causes skin irritation. May cause drowsiness or dizziness. May be fatal if swallowed and enters airways. Highly flammable liquid and vapour. Very toxic to aquatic life with long lasting effects.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. Avoid release to the environment. IF ON SKIN: Wash with plenty of soap and water. Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

Danger