Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Heptane, HPLC Grade, J.T.Baker™

High purity solvents for use in analytical and preparative separations

Manufacturer:  J.T.Baker 917733

Quantity 4L
Packaging Narrow Mouth PVC Coated Amber Glass Bottle

 View more versions of this product

Catalog No. 02-002-982



Add to cart

Description

Description

    For use in liquid chromatography (HPLC & UHPLC) & spectrophotometry; For laboratory, research, or manufacturing use
Specifications

Chemical Identifiers

142-82-5
100.205
n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
CHEBI:43098
CCCCCCC
C7H16
IMNFDUFMRHMDMM-UHFFFAOYSA-N
8900
heptane

Specifications

142-82-5
n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
CCCCCCC
100.205
CHEBI:43098
Meets ACS reagent chemical requirements
1L = 0.68kg
4L
C7H16
IMNFDUFMRHMDMM-UHFFFAOYSA-N
heptane
8900
100.2
Narrow Mouth PVC Coated Amber Glass Bottle
−4.00°C (−39.2.°F)
n-Heptane
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit