Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Heptyl sulfate, sodium salt, 99%, Acros Organics

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
18981-98-1
C7H16NaO4S
219.251
SLGMFWJQQBPOMZ-UHFFFAOYSA-N
heptyl sodium sulfate
131875435
heptyl hydrogen sulfate;sodium
CCCCCCCOS(=O)(=O)O.[Na]
18981-98-1
C7H16NaO4S
219.251
SLGMFWJQQBPOMZ-UHFFFAOYSA-N
heptyl sodium sulfate
131875435
heptyl hydrogen sulfate;sodium
CCCCCCCOS(=O)(=O)O.[Na]
Quantity 1g
Packaging Glass bottle
Assay Percent Range 99%
Formula Weight 218.25
Chemical Name or Material n-Heptyl sulfate, sodium salt