Alfa Aesar™ n-Hexadecane, 99.5+%

Manufacturer: Alfa Aesar™ 4451106

Quantity 5g

 View more versions of this product

Catalog No. AA4451106

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 544-76-3
    Molecular Formula CH3(CH2)14CH3
    Formula Weight 226.45
    MDL Number MFCD00008998
    Beilstein 1736592
    Chemical Name or Material n-Hexadecane

    Specifications

    Formula Weight 226.45
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    Specific treatment (see label).
    If skin irritation occurs: Get medical advice/attention.
    If eye irritation persists: Get medical advice/attention.
    TSCA TSCA
    Molecular Formula CH3(CH2)14CH3
    Beilstein 1736592
    Density 0.773g/mL
    Quantity 5g
    Physical Form Liquid
    MDL Number MFCD00008998
    Health Hazard 1 Warning
    RTECS Number ML9200000
    Color Colorless
    Refractive Index 1.4345
    CAS 544-76-3
    Health Hazard 2 GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.
    Melting Point 18°C
    Flash Point 135°C (275°F)
    Boiling Point 287°C
    Chemical Name or Material n-Hexadecane
    Assay ≥99.5%

    GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    Specific treatment (see label).
    If skin irritation occurs: Get medical advice/attention.
    If eye irritation persists: Get medical advice/attention.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit