Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Hexadecane, 99%, anhydrous, AcroSeal™, Acros Organics

Click to view available options
Quantity:
100mL
1L
Packaging:
AcroSeal™ Glass Bottle
544-76-3
C16H34
226.44
DCAYPVUWAIABOU-UHFFFAOYSA-N
n-hexadecane,cetane,n-cetane,hexadecan,hexadekan,cetan,zetan,unii-f8z00shp6q,ccris 5833,pentadecane, methyl
11006
CHEBI:45296
hexadecane
CCCCCCCCCCCCCCCC
544-76-3
C16H34
226.44
DCAYPVUWAIABOU-UHFFFAOYSA-N
n-hexadecane,cetane,n-cetane,hexadecan,hexadekan,cetan,zetan,unii-f8z00shp6q,ccris 5833,pentadecane, methyl
11006
CHEBI:45296
hexadecane
CCCCCCCCCCCCCCCC
Melting Point 18°C
Refractive Index 1.4330 to 1.435
Boiling Point 287°C
Flash Point 135°C
Specific Gravity 0.773
Viscosity 3.032 mPa.s (25°C)
Infrared Spectrum Authentic
Density 0.7730g/mL
Color Colorless
Quantity 100mL
Water 0.005% max.
Packaging AcroSeal™ Glass Bottle
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
Linear Formula CH3(CH2)14CH3
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,acetone and ether
Formula Weight 226.44
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material n-Hexadecane
Show More Show Less

GHS H Statement:
May be fatal if swallowed and enters airways.
Repeated exposure may cause skin dryness or cracking.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 208-878-9