Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Hexadecyl mercaptan, 90%, technical, ACROS Organics™

Click to view available options
Quantity:
10mL
100mL
500mL
Packaging:
Glass bottle
2917-26-2
C16H34S
258.5
MFCD00011677
ORTRWBYBJVGVQC-UHFFFAOYSA-N
1-hexadecanethiol,hexadecanethiol,n-hexadecanethiol,cetyl mercaptan,n-hexadecyl mercaptan,hexadecyl mercaptan,unii-qr98qio1ql,1-cetanethiol,qr98qio1ql,cetylmercaptan
18015
hexadecane-1-thiol
CCCCCCCCCCCCCCCCS
2917-26-2
C16H34S
258.5
MFCD00011677
ORTRWBYBJVGVQC-UHFFFAOYSA-N
1-hexadecanethiol,hexadecanethiol,n-hexadecanethiol,cetyl mercaptan,n-hexadecyl mercaptan,hexadecyl mercaptan,unii-qr98qio1ql,1-cetanethiol,qr98qio1ql,cetylmercaptan
18015
hexadecane-1-thiol
CCCCCCCCCCCCCCCCS
Specific Gravity 0.84
Melting Point 18°C to 20°C
Flash Point 101°C
Refractive Index 1.4610 to 1.463
Boiling Point 184°C to 191°C (7.0mmHg)
Quantity 500mL
Infrared Spectrum Authentic
Color Undesignated
Density 0.8400g/mL
Packaging Glass bottle
Assay Percent Range 88% min. (GC)
Linear Formula CH3(CH2)15SH
Beilstein 01, 430
Solubility Information Solubility in water: insoluble in water. Other solubilities: slightly soluble in ethyl alcohol,carbon tetrachl,soluble in ether
Formula Weight 258.5
Physical Form Liquid or Waxy Solid
Percent Purity 90%
Grade Technical
Chemical Name or Material n-Hexadecyl mercaptan
Show More Show Less

EINECSNumber : 220-846-6