Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Hexadecyl palmitate, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
540-10-3
C32H64O2
480.86
MFCD00053739
PXDJXZJSCPSGGI-UHFFFAOYSA-N
cetyl palmitate,hexadecyl palmitate,palmityl palmitate,cetin,hexadecanoic acid, hexadecyl ester,standamul 1616,hexadecanoic acid hexadecyl ester,hexadecanyl hexadecanoate,n-hexadecyl hexadecanoate,palmitic acid palmityl ester
10889
CHEBI:75584
hexadecyl hexadecanoate
CCCCCCCCCCCCCCCCOC(=O)CCCCCCCCCCCCCCC
540-10-3
C32H64O2
480.86
MFCD00053739
PXDJXZJSCPSGGI-UHFFFAOYSA-N
cetyl palmitate,hexadecyl palmitate,palmityl palmitate,cetin,hexadecanoic acid, hexadecyl ester,standamul 1616,hexadecanoic acid hexadecyl ester,hexadecanyl hexadecanoate,n-hexadecyl hexadecanoate,palmitic acid palmityl ester
10889
CHEBI:75584
hexadecyl hexadecanoate
CCCCCCCCCCCCCCCCOC(=O)CCCCCCCCCCCCCCC
Boiling Point 360°C
Quantity 25g
Packaging Glass bottle
Color White
Infrared Spectrum Authentic
Melting Point 52°C to 56°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula CH3(CH2)14COO(CH2)15CH3
Merck Index 15, 2030
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether
Formula Weight 480.86
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material n-Hexadecyl palmitate
Show More Show Less

EINECSNumber : 208-736-6