Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Hexadecyl sulfate, sodium salt +98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
1120-01-0
C16H33NaO4S
344.486
MFCD00047766
GGHPAKFFUZUEKL-UHFFFAOYSA-M
sodium hexadecyl sulfate,sodium cetyl sulfate,sodium n-hexadecyl sulfate,unii-3v3y3o7biq,conco sulfate c,avitex c,avitex sf,sodium hexyldecyl sulfate,tergitol anionic 7,3v3y3o7biq
23695542
sodium;hexadecyl sulfate
CCCCCCCCCCCCCCCCOS(=O)(=O)[O-].[Na+]
1120-01-0
C16H33NaO4S
344.486
MFCD00047766
GGHPAKFFUZUEKL-UHFFFAOYSA-M
sodium hexadecyl sulfate,sodium cetyl sulfate,sodium n-hexadecyl sulfate,unii-3v3y3o7biq,conco sulfate c,avitex c,avitex sf,sodium hexyldecyl sulfate,tergitol anionic 7,3v3y3o7biq
23695542
sodium;hexadecyl sulfate
CCCCCCCCCCCCCCCCOS(=O)(=O)[O-].[Na+]
Color White
Quantity 5g
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98+%
Linear Formula CH3(CH2)15OSO3Na
Formula Weight 344.49
Physical Form Powder
Percent Purity ≥98%
Chemical Name or Material n-Hexadecyl sulfate, sodium salt
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.