Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Hexane, 97%, Extra Dry over Molecular Sieve, AcroSeal™, ACROS Organics™

Click to view available options
Quantity:
100mL
1L
Packaging:
AcroSeal™ Glass Bottle
110-54-3
C6H14
86.178
MFCD00009520
VLKZOEOYAKHREP-UHFFFAOYSA-N
n-hexane,esani,skellysolve b,gettysolve-b,hexyl hydride,hexan,dipropyl,heksan,hexanen,hexanes
8058
CHEBI:29021
hexane
CCCCCC
110-54-3
C6H14
86.178
MFCD00009520
VLKZOEOYAKHREP-UHFFFAOYSA-N
n-hexane,esani,skellysolve b,gettysolve-b,hexyl hydride,hexan,dipropyl,heksan,hexanen,hexanes
8058
CHEBI:29021
hexane
CCCCCC
CAS Min % 96.0
CAS Max % 100.0
Acetic Acid 0.001% max.
Assay Percent Range 97%
Linear Formula CH3(CH2)4CH3
Beilstein 01, 142
Merck Index 15, 4732
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol, acetone, ether and chloroform
Formula Weight 86.18
Physical Form Liquid
Percent Purity 97%
Grade Extra Dry over Molecular Sieve
Density 0.6590g/mL
Packaging AcroSeal™ Glass Bottle
Refractive Index 1.3748 to 1.3810
Solubility Solubility in water: insoluble. Other solubilities: soluble in alcohol, acetone, ether and chloroform
Specific Gravity 0.659
Water 0.005% Max. (Coulometric)
Flash Point −22°C
Viscosity 0.31 mPa.s (20°C)
Boiling Point 69.0°C
Color Colorless
Melting Point -95.0°C
Quantity 100mL
Chemical Name or Material n-Hexane, Over Molecular Sieves
Show More Show Less

GHS P Statement May be fatal if swallowed and enters airways. May cause drowsiness or dizziness. Causes skin irritation. May cause damage to organs through prolonged or repeated exposure. Toxic to aquatic life with long lasting effects. Suspected of damaging fertility. Highly flammable liquid and vapor.

GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Use personal protective equipment as required. Do not breathe dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. Avoid release to the environment. IF ON SKIN: Wash with plenty of soap and water.

Danger

EINECSNumber : 203-777-6

RTECSNumber : MN9275000

TSCA : TSCA