Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Hexane, Alfa Aesar™

Click to view available options
Quantity:
1L
4L
4 x 1L
110-54-3
C6H14
86.178
MFCD00009520
VLKZOEOYAKHREP-UHFFFAOYSA-N
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
8058
CHEBI:29021
hexane
CCCCCC
110-54-3
C6H14
86.178
MFCD00009520
VLKZOEOYAKHREP-UHFFFAOYSA-N
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
8058
CHEBI:29021
hexane
CCCCCC
Assay Percent Range 95% min
Beilstein 1730733
Solubility Information Very soluble in ethanol,ethyl ether and chloroform.
Formula Weight 86.18
Physical Form Liquid, Distilled in Glass
Grade HPLC
Density 0.659
Refractive Index 1.375
Flash Point -23°C (-9°F)
Boiling Point 69°C
Melting Point °C to 95°C
Quantity 1L
Chemical Name or Material n-Hexane
Show More Show Less

P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280i-P281-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P331-P332+P313-P363-P370+P378q-P501c

H225-H304-H315-H319-H335-H336-H361f-H372

DOTInformation : Hazard Class: 3; Packaging Group: II

EINECSNumber : 203-777-6

RTECSNumber : MN9275000

TSCA : Yes

Recommended Storage : Ambient temperatures