Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Hexane, Environmental Grade, >95%, Alfa Aesar™

Click to view available options
Quantity:
1L
4L
4 x 1L
4 x 4L
110-54-3
C6H14
86.178
MFCD00009520
VLKZOEOYAKHREP-UHFFFAOYSA-N
n-hexane,esani,skellysolve b,gettysolve-b,hexyl hydride,hexan,dipropyl,heksan,hexanen,hexanes
8058
CHEBI:29021
hexane
CCCCCC
110-54-3
C6H14
86.178
MFCD00009520
VLKZOEOYAKHREP-UHFFFAOYSA-N
n-hexane,esani,skellysolve b,gettysolve-b,hexyl hydride,hexan,dipropyl,heksan,hexanen,hexanes
8058
CHEBI:29021
hexane
CCCCCC

For Pesticide Residue, Environmental and GC Analysis

Assay Percent Range >95%
Appearance Colorless liquid
Beilstein 1730733
Solubility Information Very soluble in ethanol,ethyl ether and chloroform.
Formula Weight 86.18
Physical Form Liquid
Grade Environmental
Density 0.659 g/cm3 at 20°C
Ignition Point 240°C (464°F)
Odor Weak, characteristic
Refractive Index 1.375
Flash Point -23°C (-9F)
Boiling Point 69°C
Color Colorless
Melting Point °C to 95°C
Quantity 4L
Chemical Name or Material n-Hexane
Show More Show Less

P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280i-P281-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P331-P332+P313-P363-P370+P378q-P501c

H225-H304-H315-H319-H335-H336-H361f-H372

DOTInformation : DOT Class: 3, Packaging Group: II

EINECSNumber : 203-777-6

RTECSNumber : MN9275000

TSCA : Yes

Recommended Storage : Ambient temperatures