n-Hexane, Spectrophotometric Grade, >95%, Alfa Aesar™

Manufacturer: Alfa Aesar 32454M1

Quantity 4 x 1L

 View more versions of this product

Catalog No. AA32454M1

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 110-54-3
    Physical Form Liquid
    Assay Percent Range >95%
    Molecular Formula CH3(CH2)4CH3
    Formula Weight 86.18
    MDL Number MFCD00009520
    Chemical Name or Material n-Hexane
    Grade Spectrophotometric grade

    Specifications

    Formula Weight 86.18
    Quantity 4 x 1L
    Color Colorless
    UN Number 1208; Colorless
    DOT Information DOT Class: 3; Packing Group: II
    EINECS Number 203-777-6
    Physical Form Liquid
    Odor Weak, characteristic
    Vapor Pressure 160 hPa (120mm Hg) at 20°C
    Molecular Formula CH3(CH2)4CH3
    CAS 110-54-3
    Appearance Colorless liquid
    Assay Percent Range >95%
    MDL Number MFCD00009520
    Health Hazard 1 H225-H304-H361-H373-H315-H336-H411: Highly flammable liquid and vapour. May be fatal if swallowed and enters airways. Suspected of damaging fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. Causes skin irritation. May cause drowsiness or dizziness. Toxic to aquatic life with long lasting effects.
    Chemical Name or Material n-Hexane
    Grade Spectrophotometric grade

    H225-H304-H361-H373-H315-H336-H411: Highly flammable liquid and vapour. May be fatal if swallowed and enters airways. Suspected of damaging fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. Causes skin irritation. May cause drowsiness or dizziness. Toxic to aquatic life with long lasting effects.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit