Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Hexyl Sulfide 95%, ACROS Organics™

Click to view available options
Quantity:
25g
Packaging:
Glass bottle
6294-31-1
C12H26S
202.4
MFCD00009528
LHNRHYOMDUJLLM-UHFFFAOYSA-N
hexyl sulfide,hexane, 1,1'-thiobis,dihexyl sulfide,di-n-hexyl sulfide,7-thiatridecane,dihexyl sulphide,n-hexyl sulfide,di-n-hexylsulfide,hexylthiohexane,normal-hexyl sulfide
80517
1-hexylsulfanylhexane
CCCCCCSCCCCCC
6294-31-1
C12H26S
202.4
MFCD00009528
LHNRHYOMDUJLLM-UHFFFAOYSA-N
hexyl sulfide,hexane, 1,1'-thiobis,dihexyl sulfide,di-n-hexyl sulfide,7-thiatridecane,dihexyl sulphide,n-hexyl sulfide,di-n-hexylsulfide,hexylthiohexane,normal-hexyl sulfide
80517
1-hexylsulfanylhexane
CCCCCCSCCCCCC
Infrared Spectrum Authentic
Color Colorless
Density 0.8400g/mL
Quantity 25g
Refractive Index 1.4560 to 1.4600 (20°C, 589nm)
Packaging Glass bottle
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Linear Formula [CH3(CH2)5]2S
Beilstein 01, 409
Formula Weight 202.4
Physical Form Liquid
Percent Purity ≥94%
Chemical Name or Material n-Hexyl sulfide, 95%
Show More Show Less

STENCH

EINECSNumber : 228-556-1

TSCA : TSCA