Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ N-Hydroxy-1,8-naphthalimide, 97%

Click to view available options
Quantity:
5g
25g
100g
7797-81-1
C12H7NO3
213.192
MFCD00065062
KTWCUGUUDHJVIH-UHFFFAOYSA-N
n-hydroxynaphthalimide,n-hydroxy-1,8-naphthalimide,naphthalimide, n-hydroxy,naphthalhydroxamic acid,2-hydroxy-1h-benzo de isoquinoline-1,3 2h-dione,n-hydroxy-1,8-naphthalenedicarboximide,2-hydroxy-1h-benz de isoquinoline-1,3 2h-dione,2-hydroxybenzo de isoquinoline-1,3-dione,1h-benz de isoquinoline-1,3 2h-dione, 2-hydroxy,unii-dzg9avw5v2
82263
2-hydroxybenzo[de]isoquinoline-1,3-dione
C1=CC2=C3C(=C1)C(=O)N(C(=O)C3=CC=C2)O
7797-81-1
C12H7NO3
213.192
MFCD00065062
KTWCUGUUDHJVIH-UHFFFAOYSA-N
n-hydroxynaphthalimide,n-hydroxy-1,8-naphthalimide,naphthalimide, n-hydroxy,naphthalhydroxamic acid,2-hydroxy-1h-benzo de isoquinoline-1,3 2h-dione,n-hydroxy-1,8-naphthalenedicarboximide,2-hydroxy-1h-benz de isoquinoline-1,3 2h-dione,2-hydroxybenzo de isoquinoline-1,3-dione,1h-benz de isoquinoline-1,3 2h-dione, 2-hydroxy,unii-dzg9avw5v2
82263
2-hydroxybenzo[de]isoquinoline-1,3-dione
C1=CC2=C3C(=C1)C(=O)N(C(=O)C3=CC=C2)O
Applications
It is used in chemical research & chemical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
Quantity 25g
Melting Point 283°C to 286°C
Beilstein 178595
Formula Weight 213.19
Percent Purity 97%
Chemical Name or Material N-Hydroxy-1,8-naphthalimide

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA