Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Hydroxymaleimide, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
4814-74-8
C4H3NO3
113.072
MFCD00005501
BUXKULRFRATXSI-UHFFFAOYSA-N
n-hydroxymaleimide,1-hydroxy-1h-pyrrole-2,5-dione,1-hydroxy-2,5-dihydro-1h-pyrrole-2,5-dione,n-hydroxymaleamine,n-hydroxymaleinimide,acmc-20ak3w,1h-pyrrole-2,5-dione,1-hydroxy
78535
1-hydroxypyrrole-2,5-dione
C1=CC(=O)N(C1=O)O
4814-74-8
C4H3NO3
113.072
MFCD00005501
BUXKULRFRATXSI-UHFFFAOYSA-N
n-hydroxymaleimide,1-hydroxy-1h-pyrrole-2,5-dione,1-hydroxy-2,5-dihydro-1h-pyrrole-2,5-dione,n-hydroxymaleamine,n-hydroxymaleinimide,acmc-20ak3w,1h-pyrrole-2,5-dione,1-hydroxy
78535
1-hydroxypyrrole-2,5-dione
C1=CC(=O)N(C1=O)O
Coupling reagent
Infrared Spectrum Authentic
Quantity 1g
Color Yellow to Brown
Packaging Glass bottle
Melting Point 129°C to 134°C
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 113.07
Physical Form Crystalline Powder
Percent Purity ≥96.0%
Chemical Name or Material N-Hydroxymaleimide
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Harmful if inhaled.
Causes serious eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger